Tris(4-fluorophenyl)phosphine 98%

Supplier: Thermo Scientific Chemicals
Warning

A14813-03 A14813-06 A14813-14
CAAAA14813-03EA 53.72 CAD
CAAAA14813-03 CAAAA14813-06 CAAAA14813-14
Tris(4-fluorophenyl)phosphine 98%
Tris(4-fluorophenyl)phosphine

Purity: 98% CAS No.: 18437-78-0 Molecular Formula: C18H12F3P Formula Weight: 316.27 Melting Point: 78-82° Beilstein Reference: 919838

Formula: C₁₈H₁₂F₃P
MW: 316.26
Melting Pt: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00013553
CAS Number: 18437-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR