2,3-Dichlorobenzonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14809-14
A14809-06
CAAAAA14809-14EA
682.56
CAD
CAAAAA14809-14
CAAAA14809-06
2,3-Dichlorobenzonitrile 98%
2,3-Dichlorobenzonitrile
Purity: 98%
CAS No.: 6574-97-6
Molecular Formula: C7H3Cl2N
Formula Weight: 172.01
Melting Point: 60-65°
Boiling Point: 114°/6mm
UN No.: 3439
MDL No.: MFCD00016372
Formula:
C₇H₃Cl₂N MW: 172.01 g/mol Boiling Pt: 114 °C/6 mm Melting Pt: 60…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016372 CAS Number: 6574-97-6 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...