1,2-Dibromo-4,5-difluorobenzene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14804-06
A14804-14
A14804-22
CAAAA14804-06EA
99.38
CAD
CAAAA14804-06
CAAAA14804-14
CAAAA14804-22
1,2-Dibromo-4,5-difluorobenzene 98%
1,2-Dibromo-4,5-difluorobenzene
Purity: 98%
CAS No.: 64695-78-9
Molecular Formula: C6H2Br2F2
Formula Weight: 271.89
Melting Point: 31-35°
Beilstein Reference: 1941132
Formula:
C₆H₂Br₂F₂ Boiling Pt: 208 °C (1013 hPa) Melting Pt: 31…35 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009890 CAS Number: 64695-78-9 EINECS: 265-021-1 UN: 1325 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...