3,5-Dimethoxy-p-toluic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,5-Dimethoxy-4-methylbenzoic acid

A14801-22 A14801-06 A14801-14
CAAAAA14801-22EA 1186.58 CAD
CAAAAA14801-22 CAAAA14801-06 CAAAA14801-14
3,5-Dimethoxy-p-toluic acid 97%
3,5-Dimethoxy-p-toluic acid

Purity: 97% CAS No.: 61040-81-1 Molecular Formula: C10H12O4 Formula Weight: 196.20 Melting Point: 210-216° Beilstein Reference: 2695238

Formula: C₁₀H₁₂O₄
MW: 196.2 g/mol
Melting Pt: 210…216 °C
Storage Temperature: Ambient
MDL Number: MFCD00017506
CAS Number: 61040-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR