Phenyl vinyl sulfone 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14794-03
A14794-06
A14794-14
CAAAA14794-03EA
54.19
CAD
CAAAA14794-03
CAAAA14794-06
CAAAA14794-14
Phenyl vinyl sulfone 99+%
Phenyl vinyl sulphone
Purity: 99+%
CAS No.: 5535-48-8
Molecular Formula: C8H8O2S
Formula Weight: 168.22
Storage Temperature: 2°C to 8°C
Melting Point: 67-71°
Beilstein Reference: 1906894
MDL No.: MFCD00007554
Formula:
C₈H₈O₂S Melting Pt: 67…71 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007554 CAS Number: 5535-48-8 EINECS: 226-890-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...