2-Acetylnaphthalene 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2´-Acetonaphthone, 1-(Naphthalen-2-yl)ethanone, Methyl β-naphthyl ketone, 2-Acetonaphthone, 2'-Acetonaphthone

A14793-22 A14793-18 A14793-30
CAAAAA14793-22EA 76.95 CAD
CAAAAA14793-22 CAAAA14793-0B CAAAA14793-18 CAAAA14793-30
2-Acetylnaphthalene 99%
2-Acetylnaphthalene

Purity: 99% CAS No.: 93-08-3 Molecular Formula: C12H10O Formula Weight: 170.21 Flash Point: 168°(334°F) Melting Point: 52-56° Boiling Point: 300-301° Beilstein Reference: 774965 UN No.: 3077 MDL No.: MFCD00004108

Formula: C₁₂H₁₀O
MW: 170.21 g/mol
Boiling Pt: 300…301 °C
Melting Pt: 52…56 °C
Density: 1.1171 g/cm³ (20 °C)
Flash Pt: 168 °C (334 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004108
CAS Number: 93-08-3
EINECS: 202-216-2
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR