2-Fluorobenzyl alcohol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-Fluorophenyl)methanol, 2-Fluorobenzyl Alcohol

A14791-22 A14791-09 A14791-18
CAAAAA14791-22EA 339.7 CAD
CAAAAA14791-22 CAAAA14791-09 CAAAA14791-18
2-Fluorobenzyl alcohol 98%
2-Fluorobenzyl alcohol

Purity: 98% CAS No.: 446-51-5 Molecular Formula: C7H7FO Formula Weight: 126.13 Flash Point: 90°(194°F) Refractive Index: 1.5140 Odor: Characteristic Boiling Point: 201° Beilstein Reference: 2079486 MDL No.: MFCD00004601

Formula: C₇H₇FO
Boiling Pt: 201 °C
Density: 1.175
Flash Pt: 90 °C (194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004601
CAS Number: 446-51-5
EINECS: 207-170-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR