Capric acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Decylic acid, Decanoic acid

A14788-0B A14788-0I A14788-30
CAAAA14788-0BEA 221.2 CAD
CAAAA14788-0B CAAAA14788-0I CAAAA14788-30
Capric acid 99%
Capric acid

Purity: 99% CAS No.: 334-48-5 Molecular Formula: CH3(CH2)8CO2H Formula Weight: 172.27 Flash Point: 147°(296°F) Physical Form: Cryst solid Density: 0.893 Appearance: White pH: 4 Melting Point: 29-33° Boiling Point: 268-270° Beilstein Reference: 1754556 Merck Reference: 14,1758 MDL No.: MFCD00004441

Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
Boiling Pt: 268…270 °C
Melting Pt: 29…33 °C
Density: 0.893
Flash Pt: 147 °C (296 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004441
CAS Number: 334-48-5
EINECS: 206-376-4
Merck Index: 14,1758

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR