o-Tolylacetonitrile 98+%

Supplier: Thermo Scientific Chemicals
Warning

A14787-06 A14787-14 A14787-22
CAAAA14787-06EA 55.14 CAD
CAAAA14787-06 CAAAA14787-14 CAAAA14787-22
o-Tolylacetonitrile 98+%
o-Tolylacetonitrile

Purity: 98+% CAS No.: 22364-68-7 Molecular Formula: C9H9N Formula Weight: 131.18 Flash Point: 110°(230°F) Refractive Index: 1.5275 Odor: Almond-like Boiling Point: 211-213° Beilstein Reference: 907182 UN No.: 3276 MDL No.: MFCD00001904

Formula: C₉H₉N
Boiling Pt: 211…213 °C
Melting Pt: 58…59 °C
Density: 1.010
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001904
CAS Number: 22364-68-7
EINECS: 244-937-5
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR