2-Chlorobenzoyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
o-Chlorobenzoyl chloride
A14785-22
A14785-0B
A14785-30
CAAAAA14785-22EA
69.52
CAD
CAAAAA14785-22
CAAAA14785-0B
CAAAA14785-30
2-Chlorobenzoyl chloride 97%
2-Chlorobenzoyl chloride
Purity: 97%
CAS No.: 609-65-4
Molecular Formula: C7H4Cl2O
Formula Weight: 175.01
Flash Point: 125°(257°F)
Refractive Index: 1.5718
Odor: Acrid
Melting Point: -4°
Boiling Point: 237-239°
Beilstein Reference: 386435
UN No.: 3265
MDL No.: MFCD00000660
Formula:
C₇H₄Cl₂O Boiling Pt: 237…239 °C Melting Pt: –4 °C Density: 1.379 Flash Pt: 125 °C (257 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000660 CAS Number: 609-65-4 EINECS: 210-194-0 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...