1-Iodoperfluoropropane 97% stabilized
Supplier: Thermo Scientific Chemicals
Synonyms:
Perfluoropropyl iodide, Heptafluoro-1-iodopropane, 1,1,2,2,3,3,3-Heptafluoro-1-iodopropane
Stabilization:
Stabilised with copper
A14784-06
A14784-14
CAAAA14784-06EA
106.81
CAD
CAAAA14784-06
CAAAA14784-14
1-Iodoperfluoropropane 97% stabilized
1-Iodoperfluoropropane
Purity: 97%
CAS No.: 754-34-7
Molecular Formula: CF3CF2CF2I
Formula Weight: 295.93
Storage Temperature: 2°C to 8°C
Refractive Index: 1.3290
Melting Point: -95°
Boiling Point: 40-41°
Beilstein Reference: 1758822
MDL No.: MFCD00001061
Formula:
C₃F₇I MW: 295.93 g/mol Boiling Pt: 40…41 °C Melting Pt: –95 °C Density: 2.056 Storage Temperature: Refrigerator |
MDL Number:
MFCD00001061 CAS Number: 754-34-7 EINECS: 212-045-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...