2-Methylbenzophenone 98+%

Supplier: Thermo Scientific Chemicals

A14779-09 A14779-18 A14779-30
CAAAA14779-09EA 73 CAD
CAAAA14779-09 CAAAA14779-18 CAAAA14779-30
2-Methylbenzophenone 98+%
2-Methylbenzophenone

Purity: 98+% CAS No.: 131-58-8 Molecular Formula: C14H12O Formula Weight: 196.25 Flash Point: 110°(230°F) Refractive Index: 1.5950 Melting Point: -18° Boiling Point: 309-311° Beilstein Reference: 2045469 Merck Reference: 14,7317 MDL No.: MFCD00008518

Formula: C₁₄H₁₂O
Boiling Pt: 309…311 °C
Melting Pt: –18 °C
Density: 1.085
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008518
CAS Number: 131-58-8
EINECS: 205-032-0
Merck Index: 14,7317

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR