HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
HEPES free acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulfonic acid, 4-(2-Hydroxyethyl)piperazin-1-ylethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid), 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid
A zwitterionic buffer
Purity: 99%
CAS No.: 7365-45-9
Molecular Formula: C8H18N2O4S
Formula Weight: 238.31
Physical Form: Powder
Appearance: White
Melting Point: ca 236° dec.
Beilstein Reference: 883043
Merck Reference: 14,4654
MDL No.: MFCD00006158
A zwitterionic buffer
Formula:
C₈H₁₈N₂O₄S MW: 238.31 g/mol Melting Pt: ∼ 236 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006158 CAS Number: 7365-45-9 EINECS: 230-907-9 Merck Index: 14,4654 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...