3-(Methylthio)propanal 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-(Methylthio)propionaldehyde

A14774-09 A14774-18 A14774-30
CAAAA14774-09EA 71.73 CAD
CAAAA14774-09 CAAAA14774-18 CAAAA14774-30
3-(Methylthio)propanal 98%
3-(Methylthio)propanal

Purity: 98% CAS No.: 3268-49-3 Molecular Formula: C4H8OS Formula Weight: 104.17 Storage Temperature: 2°C to 8°C Flash Point: 64°(147°F) Physical Form: Liquid Density: 1.052 Refractive Index: 1.4825 Sensitivity: Air Sensitive Appearance: Colorless to pale yellow Odor: Strong Melting Point: -58° Boiling Point: 165-166° Beilstein Reference: 1739289 UN No.: 2785 MDL No.: MFCD00007022

Formula: C₄H₈OS
MW: 104.17 g/mol
Boiling Pt: 165…166 °C
Melting Pt: –58 °C
Density: 1.052
Flash Pt: 64 °C (147 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00007022
CAS Number: 3268-49-3
EINECS: 221-882-5
UN: 2785
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR