Diethyl methylphosphonate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14772-14
A14772-09
A14772-18
CAAAAA14772-14EA
387.1
CAD
CAAAAA14772-14
CAAAA14772-09
CAAAA14772-18
Diethyl methylphosphonate 97%
Diethyl methylphosphonate
Purity: 97%
CAS No.: 683-08-9
Molecular Formula: C5H13O3P
Formula Weight: 152.13
Flash Point: 75°(167°F)
Refractive Index: 1.4140
Boiling Point: 194°
Beilstein Reference: 1753416
MDL No.: MFCD00009813
Formula:
C₅H₁₃O₃P MW: 152.13 g/mol Boiling Pt: 194 °C Density: 1.040 Flash Pt: 75 °C (167 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009813 CAS Number: 683-08-9 EINECS: 211-667-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...