(3-Pyridyl)acetic acid ethyl ester 99%

Supplier: Thermo Scientific Chemicals
Warning

A14768-06 A14768-14
CAAAA14768-06EA 58.93 CAD
CAAAA14768-06 CAAAA14768-14 CAAAA14768-22
(3-Pyridyl)acetic acid ethyl ester 99%
(3-Pyridyl)acetic acid ethyl ester

Purity: 99% CAS No.: 39931-77-6 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Flash Point: 110°(230°F) Refractive Index: 1.500 Boiling Point: 79-80°/0.4mm Beilstein Reference: 123827 MDL No.: MFCD00006411

Formula: C₉H₁₁NO₂
Boiling Pt: 79…80 °C/0.4 mm
Density: 1.086
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006411
CAS Number: 39931-77-6
EINECS: 254-707-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR