Methyl-3-aminobenzoate 98+%

Supplier: Thermo Scientific Chemicals
Warning

A14766-22 A14766-06 A14766-14
CAAAAA14766-22EA 970.12 CAD
CAAAAA14766-22 CAAAA14766-06 CAAAA14766-14
Methyl-3-aminobenzoate 98+%
Methyl 3-aminobenzoate

Purity: 98+% CAS No.: 4518-10-9 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Melting Point: 49-53° Beilstein Reference: 1101468

Formula: C₈H₉NO₂
Boiling Pt: 120 °C (0.7 -rr)
Melting Pt: 49…53 °C
Density: 1.232
Storage Temperature: Ambient
MDL Number: MFCD00017102
CAS Number: 4518-10-9
EINECS: 224-842-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR