1,2-Dibromobenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14765-14
A14765-22
A14765-30
A14765-18
CAAAAA14765-14EA
133.04
CAD
CAAAAA14765-14
CAAAAA14765-22
CAAAAA14765-30
CAAAA14765-18
1,2-Dibromobenzene 98%
1,2-Dibromobenzene
Purity: 98%
CAS No.: 583-53-9
Molecular Formula: C6H4Br2
Formula Weight: 235.91
Flash Point: 91°(195°F)
Refractive Index: 1.6110
Odor: Strong
Melting Point: 4-6°
Boiling Point: 224-226°
Beilstein Reference: 970241
UN No.: 3082
MDL No.: MFCD00000057
Formula:
C₆H₄Br₂ MW: 235.91 g/mol Boiling Pt: 224…226 °C Melting Pt: 4…6 °C Density: 1.985 Flash Pt: 91 °C (195 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000057 CAS Number: 583-53-9 EINECS: 209-507-3 UN: 3082 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...