6-Methyl-2-pyridone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxy-6-methylpyridine, 6-Methylpyridin-2-ol, 6-Methylpyridin-2(1H)-one

A14764-06 A14764-14
CAAAA14764-06EA 72.05 CAD
CAAAA14764-06 CAAAA14764-14
6-Methyl-2-pyridone 98%
6-Methyl-2-pyridone

Purity: 98% CAS No.: 3279-76-3 Molecular Formula: C6H7NO Formula Weight: 109.13 Odor: Odorless Melting Point: 157-161° Beilstein Reference: 107073 MDL No.: MFCD00006279

Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 283…284 °C (1013 hPa)
Melting Pt: 157…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00006279
CAS Number: 3279-76-3
EINECS: 221-919-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR