2-Methyl-5-nitrobenzenesulfonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Nitrotoluene-2-sulphonyl chloride, 4-Nitrotoluene-2-sulfonyl chloride, 2-Methyl-5-nitrobenzene-1-sulfonyl chloride, 2-Methyl-5-nitrobenzene-1-sulphonyl chloride
A14760-06
A14760-14
CAAAAA14760-22EA
1549.54
CAD
CAAAAA14760-22
CAAAA14760-06
CAAAA14760-14
2-Methyl-5-nitrobenzenesulfonyl chloride 97%
2-Methyl-5-nitrobenzenesulphonyl chloride
Purity: 97%
CAS No.: 121-02-8
Molecular Formula: C7H6ClNO4S
Formula Weight: 235.65
Flash Point:
Formula:
C₇H₆ClNO₄S Boiling Pt: 135…137 °C/0.7 mm Melting Pt: 42…46 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051695 CAS Number: 121-02-8 EINECS: 204-444-8 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...