Picolinic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

A14749-22 A14749-36
CAAAA14749-22EA 94.64 CAD
CAAAA14749-22 CAAAA14749-36
Picolinic acid 99%
Picolinic acid

Purity: 99% CAS No.: 98-98-6 Molecular Formula: C6H5NO2 Formula Weight: 123.11 Flash Point: 139°(232°F) Odor: Odorless Beilstein Reference: 109595 Merck Reference: 14,7403 MDL No.: MFCD00006293

Formula: C₆H₅NO₂
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 135…139 °C
Density: 1.49 g/cm³ (20 °C)
Flash Pt: 139 °C(232 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006293
CAS Number: 98-98-6
EINECS: 202-719-7
Merck Index: 14,7403

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR