Methyl propiolate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl acetylenecarboxylate, Propiolic acid methyl ester, 2-Propynoic acid methyl ester

A14743-06 A14743-14 A14743-22
CAAAA14743-06EA 94.33 CAD
CAAAA14743-06 CAAAA14743-14 CAAAA14743-22
Methyl propiolate 99%
Methyl propiolate

Purity: 99% CAS No.: 922-67-8 Molecular Formula: HC≡CCO2CH3 Formula Weight: 84.07 Flash Point: 10°(50°F) Refractive Index: 1.4080 Odor: Pungent Boiling Point: 98-101° Beilstein Reference: 605462 UN No.: 3272 MDL No.: MFCD00008572

Formula: C₄H₄O₂
MW: 84.07 g/mol
Boiling Pt: 98…101 °C
Density: 1.016
Flash Pt: 10 °C (50 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008572
CAS Number: 922-67-8
EINECS: 213-083-5
UN: 3272
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR