4-Nitrobenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

A14738-0B A14738-0I A14738-30
CAAAA14738-0BEA 108.55 CAD
CAAAA14738-0B CAAAA14738-0I CAAAA14738-30
4-Nitrobenzoic acid 99%
4-Nitrobenzoic acid

Purity: 99% CAS No.: 62-23-7 Molecular Formula: C7H5NO4 Formula Weight: 167.12 Melting Point: 239-241° Beilstein Reference: 973593 Merck Reference: 14,6589 MDL No.: MFCD00007352

Formula: C₇H₅NO₄
Boiling Pt: 359 °C (1013 hPa)
Melting Pt: 239…241 °C
Density: 1.610
Storage Temperature: Ambient
MDL Number: MFCD00007352
CAS Number: 62-23-7
EINECS: 200-526-2
Merck Index: 14,6589

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR