Diethyl suberate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Diethyl octanedioate, 1,8-Octanedioic acid diethyl ester, Suberic acid diethyl ester

A14735-36 A14735-18 A14735-30
CAAAAA14735-36EA 1243.46 CAD
CAAAAA14735-36 CAAAA14735-18 CAAAA14735-30
Diethyl suberate 99%
Diethyl suberate

Purity: 99% CAS No.: 2050-23-9 Molecular Formula: C12H22O4 Formula Weight: 230.30 Flash Point: 110°(230°F) Refractive Index: 1.4320 Boiling Point: 282-283° Beilstein Reference: 1785500 MDL No.: MFCD00009217

Formula: C₁₂H₂₂O₄
MW: 230.3 g/mol
Boiling Pt: 282…283 °C
Melting Pt: 5
Density: 0.980
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009217
CAS Number: 2050-23-9
EINECS: 218-084-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR