n-Decane 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14732-0F
A14732-AE
A14732-AP
CAAAA14732-0FEA
794.74
CAD
CAAAA14732-0F
CAAAA14732-AE
CAAAA14732-AP
n-Decane 99%
n-Decane
MDL: MFCD00008954
Beilstein: 1696981
Soluble in alcohol and ethers
Purity: 99%
CAS No.: 124-18-5
Molecular Formula: C10H22
Formula Weight: 142.29
Flash Point: 46°(115°F)
Physical Form: Liquid
Density: 0.730
Refractive Index: 1.4115
Appearance: Colorless
Odor: Petrol-like
Melting Point: -30°
Boiling Point: 172-174°
Solubility: Soluble in alcohol and ethers
Beilstein Reference: 1696981
UN No.: 2247
MDL No.: MFCD00008954
Formula:
H₃C(CH₂)₈CH₃ Boiling Pt: 172…174 °C Melting Pt: –30 °C Density: 0.730 Flash Pt: 46 °C (115 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008954 CAS Number: 124-18-5 EINECS: 204-686-4 UN: 2247 ADR: 3,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...