L-Arginine hydrochloride 98+%
Supplier: Thermo Scientific Chemicals
A14730-0E
A14730-22
A14730-36
CAAAA14730-0EEA
780.52
CAD
CAAAA14730-0E
CAAAA14730-22
CAAAA14730-36
L-Arginine hydrochloride 98+%
L-Arginine hydrochloride
Purity: 98+%
CAS No.: 1119-34-2
Molecular Formula: C6H14N4O2.HCl
Formula Weight: 210.67
Melting Point: 216° dec.
Beilstein Reference: 3631658
Merck Reference: 14,780
Optical Rotation: +22° (c=5 in 5N HCl)
MDL No.: MFCD00064550
Formula:
C₆H₁₅ClN₄O₂ Melting Pt: 216 °C (dec.) Density: 1.42 Storage Temperature: Ambient |
MDL Number:
MFCD00064550 CAS Number: 1119-34-2 EINECS: 214-275-1 Merck Index: 14,780 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...