1-Bromooctane 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14722-0B
A14722-0I
A14722-30
CAAAA14722-0BEA
184.86
CAD
CAAAA14722-0B
CAAAA14722-0I
CAAAA14722-30
1-Bromooctane 98+%
1-Bromooctane
Purity: 98+%
CAS No.: 111-83-1
Molecular Formula: C8H17Br
Formula Weight: 193.13
Flash Point: 78°(172°F)
Refractive Index: 1.4518
Odor: Characteristic
Melting Point: -55°
Boiling Point: 200-202°
Beilstein Reference: 1733136
Merck Reference: 14,6764
UN No.: 3082
MDL No.: MFCD00000276
Formula:
BrH₂C(CH₂)₆CH₃ Boiling Pt: 200…202 °C Melting Pt: –55 °C Density: 1.114 Flash Pt: 78 °C (172 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000276 CAS Number: 111-83-1 EINECS: 203-912-9 UN: 3082 ADR: 9,III Merck Index: 14,6764 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...