8-Quinolinol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14720-0E
A14720-22
A14720-36
CAAAA14720-0EEA
872.16
CAD
CAAAA14720-0E
CAAAA14720-22
CAAAA14720-36
8-Quinolinol 99%
8-Quinolinol
MDL: MFCD00006807
Beilstein: 114512
Almost insoluble in water and diethyl ether. Highly soluble in ethanol, acetone, chloroform, and benzene.
Purity: 99%
CAS No.: 148-24-3
Molecular Formula: C9H7NO
Formula Weight: 145.16
Melting Point: 72-76°
Boiling Point: 267°
Solubility: Almost insoluble in water and diethyl ether. Highly soluble in ethanol, acetone, chloroform, and benzene.
Beilstein Reference: 114512
Merck Reference: 14,4843
MDL No.: MFCD00006807
Formula:
C₉H₇NO Boiling Pt: 267 °C Melting Pt: 72…76 °C Density: 1.03 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006807 CAS Number: 148-24-3 EINECS: 205-711-1 UN: 2811 ADR: 6.1,III Merck Index: 14,4843 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...