6-Aminocaproic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14719-0E
A14719-22
A14719-36
CAAAA14719-0EEA
1218.18
CAD
CAAAA14719-0E
CAAAA14719-22
CAAAA14719-36
6-Aminocaproic acid 99%
6-Aminocaproic acid
Purity: 99%
CAS No.: 60-32-2
Molecular Formula: C6H13NO2
Formula Weight: 131.18
Physical Form: Cryst powder
Appearance: White
Melting Point: 210-212°
Beilstein Reference: 906872
Merck Reference: 14,432
MDL No.: MFCD00008238
Formula:
C₆H₁₃NO₂ Boiling Pt: 257 °C (1013 hPa) Melting Pt: 210…212 °C Flash Pt: 207 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008238 CAS Number: 60-32-2 EINECS: 200-469-3 Merck Index: 14,432 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...