3,5-Di-tert-butyl-4-hydroxybenzyl alcohol 97%

Supplier: Thermo Scientific Chemicals
Warning

A14709-06 A14709-14 A14709-22
CAAAA14709-06EA 83.11 CAD
CAAAA14709-06 CAAAA14709-14 CAAAA14709-22
3,5-Di-tert-butyl-4-hydroxybenzyl alcohol 97%
3,5-Di-tert-butyl-4-hydroxybenzyl alcohol

Purity: 97% CAS No.: 88-26-6 Molecular Formula: C15H24O2 Formula Weight: 236.36 Melting Point: 138-141° Boiling Point: 214°/40mm MDL No.: MFCD00017254

Formula: C₁₅H₂₄O₂
MW: 236.35 g/mol
Boiling Pt: 214 °C/40 mm
Melting Pt: 138…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00017254
CAS Number: 88-26-6
EINECS: 201-815-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR