Di-tert-butyl pyrocarbonate 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Boc Anhydride, Di-tert-butyl dicarbonate, DIBOC
Reagent for the introduction of the Boc protecting group
Purity: 97+%
CAS No.: 24424-99-5
Molecular Formula: C10H18O5
Formula Weight: 218.25
Storage Temperature: 2°C to 8°C
Flash Point: 37°(99°F)
Physical Form: Low-melting solid
Density: 0.950
Refractive Index: 1.4090
Sensitivity: Moisture Sensitive
Appearance: White to pale yellow
Odor: Mild
Melting Point: 21-24°
Boiling Point: 56-57°/0.5mm
Beilstein Reference: 1911173
UN No.: 2930
MDL No.: MFCD00008805
Reagent for the introduction of the Boc protecting group
Formula:
C₁₀H₁₈O₅ Boiling Pt: 56…57 °C/0.5 mm Melting Pt: 21…24 °C Density: 0.950 Flash Pt: 37 °C (99 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008805 CAS Number: 24424-99-5 EINECS: 246-240-1 UN: 2930 ADR: 6.1,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...