2-Bromo-5-nitrothiazole 98%

Supplier: Thermo Scientific Chemicals
Warning

A14701-06 A14701-14
CAAAA14701-03EA 94.15 CAD
CAAAA14701-03 CAAAA14701-06 CAAAA14701-14
2-Bromo-5-nitrothiazole 98%
2-Bromo-5-nitrothiazole

Purity: 98% CAS No.: 3034-48-8 Molecular Formula: C3HBrN2O2S Formula Weight: 209.03 Melting Point: 85-90° Beilstein Reference: 4856

Formula: C₃HBrN₂O₂S
Melting Pt: 85…90 °C
Density: 2,295 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005317
CAS Number: 3034-48-8
EINECS: 221-226-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR