4-Pyridylmethanol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14698-14
A14698-22
CAAAA14698-14EA
92.75
CAD
CAAAA14698-14
CAAAA14698-22
CAAAA14698-36
4-Pyridylmethanol 99%
4-Pyridylmethanol
Purity: 99%
CAS No.: 586-95-8
Molecular Formula: C6H7NO
Formula Weight: 109.13
Storage Temperature: 2°C to 8°C
Flash Point: 148°(298°F)
Melting Point: 53-55°
Boiling Point: 141°/12mm
Beilstein Reference: 107850
MDL No.: MFCD00006442
Formula:
C₆H₇NO Boiling Pt: 141 °C (12 mmHg) Melting Pt: 53…55 °C Density: 1,15 g/cm³ (20 °C) Flash Pt: 148 °C (298 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00006442 CAS Number: 586-95-8 EINECS: 209-590-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...