Cyclopentane 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14697-0F
A14697-AE
A14697-AP
CAAAA14697-0FEA
774.2
CAD
CAAAA14697-0F
CAAAA14697-AE
CAAAA14697-AP
Cyclopentane 95%
Cyclopentane
CAS No.: 287-92-3
Molecular Formula: C5H10
Formula Weight: 70.14
Flash Point: -37°(-34°F)
Physical Form: Liquid
Density: 0.748
Refractive Index: 1.4060
Appearance: Colorless
Odor: Petrol-like
Melting Point: -95°
Boiling Point: 47-49°
Beilstein Reference: 1900195
Merck Reference: 14,2741
UN No.: 1146
MDL No.: MFCD00001356
Formula:
C₅H₁₀ Boiling Pt: 47…49 °C Melting Pt: –95 °C Density: 0.748 Flash Pt: –37 °C (–34 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001356 CAS Number: 287-92-3 EINECS: 206-016-6 UN: 1146 ADR: 3,II Merck Index: 14,2741 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...