α,4-Dichlorotoluene 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14696-14
A14696-0B
A14696-0I
A14696-30
CAAAAA14696-14EA
38.24
CAD
CAAAAA14696-14
CAAAA14696-0B
CAAAA14696-0I
CAAAA14696-30
α,4-Dichlorotoluene 98+%
α,4-Dichlorotoluene
Purity: 98+%
CAS No.: 104-83-6
Molecular Formula: C7H6Cl2
Formula Weight: 161.03
Flash Point: 107°(225°F)
Refractive Index: 1.5575
Odor: Pungent
Melting Point: 26-30°
Boiling Point: 220-221°
Beilstein Reference: 471558
UN No.: 3427
MDL No.: MFCD00000914
Formula:
C₇H₆Cl₂ Boiling Pt: 220…221 °C Melting Pt: 26…30 °C Density: 1.275 Flash Pt: 107 °C (225 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000914 CAS Number: 104-83-6 EINECS: 203-242-7 UN: 3427 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...