Tricine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid, Tricine buffer, N-(Tris(hydroxymethyl)methyl)glycine
A14695-36
A14695-14
A14695-22
CAAAAA14695-36EA
420.28
CAD
CAAAAA14695-36
CAAAA14695-14
CAAAA14695-22
Tricine 98+%
Tricine
Good's buffers
Purity: 98+%
CAS No.: 5704-04-1
Molecular Formula: (HOCH2)3CNHCH2CO2H
Formula Weight: 179.17
Melting Point: ca 183° dec.
Beilstein Reference: 1937804
Merck Reference: 14,9651
MDL No.: MFCD00004277
Formula:
(HOCH₂)₃CNHCH₂CO₂H Melting Pt: ∼ 183 °C (dec.) Density: 1.523 g/cm³ (19.85 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004277 CAS Number: 5704-04-1 EINECS: 227-193-6 Merck Index: 14,9651 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...