Cyclohexanecarboxylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14693-36
A14693-30
CAAAAA14693-36EA
120.55
CAD
CAAAAA14693-36
CAAAA14693-30
Cyclohexanecarboxylic acid 98%
Cyclohexanecarboxylic acid
Purity: 98%
CAS No.: 98-89-5
Molecular Formula: C7H12O2
Formula Weight: 128.17
Flash Point: 118°(244°F)
Refractive Index: 1.4610
Melting Point: 29-32°
Boiling Point: 231-233°
Beilstein Reference: 970529
Merck Reference: 14,2724
MDL No.: MFCD00001461
Formula:
C₇H₁₂O₂ Boiling Pt: 231…233 °C Melting Pt: 29…32 °C Density: 1.048 Flash Pt: 118 °C(244 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001461 CAS Number: 98-89-5 EINECS: 202-711-3 Merck Index: 14,2724 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...