1,1'-Carbonyldiimidazole 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Di(1H-imidazol-1-yl)methanone, CDI Crosslinker, 1,1'-Carbonylbis(imidazole), 1,1'-Carbonylbis-1H-imidazole, N,N'-Carbonyldiimidazole, CDI
Peptide coupling reagent
Purity: 97%
CAS No.: 530-62-1
Molecular Formula: C7H6N4O
Formula Weight: 162.15
Physical Form: Cryst powder
Sensitivity: Moisture Sensitive
Appearance: White to cream
pH: 7.7
Melting Point: 116-120°
Beilstein Reference: 6826
Merck Reference: 14,1819
UN No.: 3263
MDL No.: MFCD00005286
Peptide coupling reagent
Formula:
C₇H₆N₄O MW: 162.15 g/mol Melting Pt: 116…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005286 CAS Number: 530-62-1 EINECS: 208-488-9 UN: 3259 ADR: 8,III Merck Index: 14,1819 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...