1,3-Dibromopropane 98%

Supplier: Thermo Scientific Chemicals
Warning

A14687-0B A14687-0I A14687-30
CAAAA14687-0BEA 352.34 CAD
CAAAA14687-0B CAAAA14687-0I CAAAA14687-30
1,3-Dibromopropane 98%
1,3-Dibromopropane

Purity: 98% CAS No.: 109-64-8 Molecular Formula: C3H6Br2 Formula Weight: 201.90 Flash Point: 54°(130°F) Physical Form: Liquid Density: 1.99 Refractive Index: 1.5230 Appearance: Colorless Melting Point: -34° Boiling Point: 167° Beilstein Reference: 635662 Merck Reference: 14,9713 UN No.: 1993 MDL No.: MFCD00000255

Formula: BrH₂CCH₂CH₂Br
MW: 201.89 g/mol
Boiling Pt: 167 °C
Melting Pt: –34 °C
Density: 1.99
Flash Pt: 54 °C (130 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000255
CAS Number: 109-64-8
EINECS: 203-690-3
UN: 1993
ADR: 3,III
Merck Index: 14,9713

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR