4-Fluorobenzyl chloride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Fluorobenzyl chloride, p-Fluorobenzyl chloride, 1-Chloromethyl-4-fluorobenzene, α-Chloro-4-fluorotoluene, 1-(Chloromethyl)-4-fluorobenzene
A14686-14
A14686-22
A14686-36
CAAAA14686-14EA
56.72
CAD
CAAAA14686-14
CAAAA14686-22
CAAAA14686-36
4-Fluorobenzyl chloride 99%
4-Fluorobenzyl chloride
Purity: 99%
CAS No.: 352-11-4
Molecular Formula: C7H6ClF
Formula Weight: 144.58
Flash Point: 69°(156°F)
Refractive Index: 1.5130
Odor: Pungent
Melting Point: -18°
Boiling Point: 185°
Beilstein Reference: 742272
UN No.: 3265
MDL No.: MFCD00000913
Formula:
C₇H₆ClF Boiling Pt: 185 °C Melting Pt: –18 °C Density: 1.214 Flash Pt: 69 °C (156 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000913 CAS Number: 352-11-4 EINECS: 206-516-4 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...