6-Benzylaminopurine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14678-03
A14678-06
A14678-14
CAAAA14678-03EA
41.08
CAD
CAAAA14678-03
CAAAA14678-06
CAAAA14678-14
6-Benzylaminopurine 99%
6-Benzylaminopurine
Inhibitor of respiratory kinase in plants. Increases post-harvest life of green vegetables
Purity: 99%
CAS No.: 1214-39-7
Molecular Formula: C12H11N5
Formula Weight: 225.26
Melting Point: 229-233°
Beilstein Reference: 19406
Inhibitor of respiratory kinase in plants. Increases post-harvest life of green vegetables
Formula:
C₁₂H₁₁N₅ Melting Pt: 229…233 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005572 CAS Number: 1214-39-7 EINECS: 214-927-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...