5-Bromo-2-thiophenesulfonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

A14677-06 A14677-14
CAAAA14677-06EA 159.58 CAD
CAAAA14677-06 CAAAA14677-14
5-Bromo-2-thiophenesulfonyl chloride 98%
5-Bromo-2-thiophenesulphonyl chloride

Purity: 98% CAS No.: 55854-46-1 Molecular Formula: C4H2BrClO2S2 Formula Weight: 261.55 Melting Point: 39-42° Boiling Point: 99-101°/0.5mm UN No.: 3261 MDL No.: MFCD00084909

Formula: C₄H₂BrClO₂S₂
Boiling Pt: 99…101 °C/0.5 mm
Melting Pt: 39…42 °C
Storage Temperature: Ambient
MDL Number: MFCD00084909
CAS Number: 55854-46-1
UN: 3261
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR