1-Bromo-4-octylbenzene 97%

Supplier: Thermo Scientific Chemicals

A14676-03 A14676-06
CAAAA14676-03EA 83.27 CAD
CAAAA14676-03 CAAAA14676-06
1-Bromo-4-octylbenzene 97%
1-Bromo-4-octylbenzene

Purity: 97% CAS No.: 51554-93-9 Molecular Formula: C14H21Br Formula Weight: 269.23 MDL No.: MFCD00038348

Formula: C₁₄H₂₁Br
Storage Temperature: Ambient
MDL Number: MFCD00038348
CAS Number: 51554-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR