n-Tetradecane 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tetradecane

A14672-0E A14672-22 A14672-36
CAAAA14672-0EEA 1178.68 CAD
CAAAA14672-0E CAAAA14672-22 CAAAA14672-36
n-Tetradecane 99%
n-Tetradecane

Purity: 99% CAS No.: 629-59-4 Molecular Formula: C14H30 Formula Weight: 198.39 Flash Point: 99°(211°F) Physical Form: Liquid Density: 0.763 Refractive Index: 1.4290 Appearance: Colorless Odor: Mild Melting Point: 6° Boiling Point: 252-254° MDL No.: MFCD00008986

Formula: H₃C(CH₂)₁₂CH₃
MW: 198.39 g/mol
Boiling Pt: 252…254 °C
Melting Pt: 6 °C
Density: 0.763
Flash Pt: 99 °C(211 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008986
CAS Number: 629-59-4
EINECS: 211-096-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03064a29015d1e810f1e4350a7a6cd2dfe4713fd&vid=f4838a5686fe033622495a3bf280963555654d9a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR