2-Isopropylaniline 97%

Supplier: Thermo Scientific Chemicals
Warning

A14671-14 A14671-22 A14671-36
CAAAA14671-14EA 59.72 CAD
CAAAA14671-14 CAAAA14671-22 CAAAA14671-36
2-Isopropylaniline 97%
2-Isopropylaniline

Purity: 97% CAS No.: 643-28-7 Molecular Formula: C9H13N Formula Weight: 135.21 Flash Point: 95°(203°F) Refractive Index: 1.5485 Boiling Point: 214-216° Beilstein Reference: 636283 MDL No.: MFCD00007720

Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 214…216 °C
Density: 0.966
Flash Pt: 95 °C (203 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007720
CAS Number: 643-28-7
EINECS: 211-397-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR