Diethylene glycol monoethyl ether 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(2-Ethoxyethoxy)ethanol, Di(ethylene glycol) ethyl ether, Diethylene Glycol Monoethyl Ether

A14670-0C A14670-0E A14670-36
CAAAA14670-0CEA 292.3 CAD
CAAAA14670-0C CAAAA14670-0E CAAAA14670-36
Diethylene glycol monoethyl ether 98%
Diethylene glycol monoethyl ether

Purity: 98% CAS No.: 111-90-0 Molecular Formula: C6H14O3 Formula Weight: 134.18 Flash Point: 96°(204°F) Refractive Index: 1.4280 Odor: Pleasant Melting Point: -80° Boiling Point: 201-202° Beilstein Reference: 1736441 Merck Reference: 14,1800 MDL No.: MFCD00002872

Formula: C₆H₁₄O₃
MW: 134.18 g/mol
Boiling Pt: 197
Melting Pt: -80
Density: 0.989
Flash Pt: 96 °C (204 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002872
CAS Number: 111-90-0
EINECS: 203-919-7
REACH: 01-2119475105-42
Merck Index: 14,1800

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR