Allyltrimethylsilane 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14662-30
A14662-09
A14662-18
CAAAAA14662-30EA
1036.48
CAD
CAAAAA14662-30
CAAAA14662-09
CAAAA14662-18
Allyltrimethylsilane 98+%
Allyltrimethylsilane
Purity: 98+%
CAS No.: 762-72-1
Molecular Formula: CH2=CHCH2Si(CH3)3
Formula Weight: 114.27
Flash Point: -10°(14°F)
Refractive Index: 1.4080
Boiling Point: 85-86°
Beilstein Reference: 906755
UN No.: 1993
MDL No.: MFCD00008635
Formula:
H₂CCHCH₂Si(CH₃)₃ Boiling Pt: 85…86 °C Density: 0.717 Flash Pt: –10 °C (14 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008635 CAS Number: 762-72-1 EINECS: 212-104-5 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...