2-Iodoethanol 99% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with copper
A14658-06
A14658-14
A14658-22
CAAAA14658-06EA
67.31
CAD
CAAAA14658-06
CAAAA14658-14
CAAAA14658-22
2-Iodoethanol 99% stabilized
2-Iodoethanol
Purity: 99%
CAS No.: 624-76-0
Molecular Formula: C2H5IO
Formula Weight: 171.96
Storage Temperature: 2°C to 8°C
Flash Point: 65°(149°F)
Refractive Index: 1.5720
Odor: Characteristic
Boiling Point: 85°/25mm
Beilstein Reference: 1098258
UN No.: 2810
MDL No.: MFCD00002830
Formula:
C₂H₅IO Boiling Pt: 85 °C/25 mm Density: 2.202 Flash Pt: 65 °C (149 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002830 CAS Number: 624-76-0 EINECS: 210-861-6 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...