2,4-Dimethylbenzaldehyde 90+%
Supplier: Thermo Scientific Chemicals
Warning
|
A14655-06
A14655-14
CAAAA14655-06EA
74.26
CAD
CAAAA14655-06
CAAAA14655-14
2,4-Dimethylbenzaldehyde 90+%
2,4-Dimethylbenzaldehyde
Purity: 90+%
CAS No.: 15764-16-6
Molecular Formula: C9H10O
Formula Weight: 134.18
Flash Point: 88°(190°F)
Refractive Index: 1.5490
Odor: Almond-like
Melting Point: -9°
Boiling Point: 215-216°
MDL No.: MFCD00003340
Formula:
C₉H₁₀O MW: 134.18 g/mol Boiling Pt: 215…216 °C Melting Pt: –9 °C Density: 1.02 Flash Pt: 88 °C(190 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003340 CAS Number: 15764-16-6 EINECS: 239-856-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...