2-Fluoro-5-nitrotoluene 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Fluoro-2-methyl-4-nitrobenzene

A14649-09 A14649-18
CAAAA14649-09EA 91.64 CAD
CAAAA14649-09 CAAAA14649-18
2-Fluoro-5-nitrotoluene 98+%
2-Fluoro-5-nitrotoluene

Purity: 98+% CAS No.: 455-88-9 Molecular Formula: C7H6FNO2 Formula Weight: 155.13 Flash Point: 105°(221°F) Melting Point: 37-40° Boiling Point: 98-101°/13mm Beilstein Reference: 1940341 UN No.: 2811 MDL No.: MFCD00007284

Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 98…101 °C/13 mm
Melting Pt: 37…40 °C
Flash Pt: 105 °C(221 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007284
CAS Number: 455-88-9
EINECS: 207-251-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR